siRNA Details
| Gene Symbol: Kcnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC139325.1 | - | 1 | 304 | 8862347 | |
| BC139327.1 | - | 1 | 304 | 8862349 | ||
| M64226.1 | - | 1 | 220 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnd1| Gene Symbol: Kcnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC139325.1 | - | 1 | 304 | 8862347 | |
| BC139327.1 | - | 1 | 304 | 8862349 | ||
| M64226.1 | - | 1 | 220 | |||