siRNA Details
| Gene Symbol: Kin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025280.2 | NP_079556.1 | 5 | 1155 | ||
| GenBank mRNA | AK132548.1 | - | 5 | 1699 | ||
| AK167740.1 | - | 5 | 1159 | |||
| BC028860.1 | - | 5 | 1155 | 3673830 | ||
| BC058169.1 | - | 4 | 2538 | |||
| X58472.1 | - | 5 | 1160 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BC099685.1 | - | 5401 | Chr. - - [-] on Build | ||

Kin
Genomic Map