siRNA Details
| Gene Symbol: Rac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133223.4 | NP_573486.1 | 2 | 223 | ||
| GenBank mRNA | AB040819.1 | - | 2 | 90 | ||
| AK089930.1 | - | 2 | 224 | |||
| AK150203.1 | - | 2 | 228 | |||
| AY974601.1 | - | 1 | 62 | |||
| BC117028.1 | - | 2 | 221 | 40126347 | ||
| BC119041.1 | - | 2 | 221 | 8733790 | ||

Rac3
Genomic Map