siRNA Details
| Gene Symbol: Pdzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133226.2 | NP_573489.2 | 10 | 1316 | ||
| GenBank mRNA | AF334612.1 | - | 10 | 1312 | ||
| AK052748.1 | - | 10 | 1317 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdzd3| Gene Symbol: Pdzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133226.2 | NP_573489.2 | 10 | 1316 | ||
| GenBank mRNA | AF334612.1 | - | 10 | 1312 | ||
| AK052748.1 | - | 10 | 1317 | |||