siRNA Details
| Gene Symbol: Tm4sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008536.3 | NP_032562.1 | 6, 7 | 1189 | ||
| GenBank mRNA | AK077326.1 | - | 4, 5 | 692 | ||
| AK133556.1 | - | 690 | ||||
| AK134603.1 | - | 5, 6 | 1126 | |||
| AK164718.1 | - | 4, 5 | 693 | |||
| BC083073.1 | - | 6, 7 | 1189 | 6337376 | ||
| KC508089.1 | - | 4, 5 | 664 | |||
| L15429.1 | - | 4, 5 | 679 | |||
| L15443.1 | - | 4, 5 | 679 | |||

Tm4sf1
Genomic Map