siRNA Details
| Gene Symbol: Vmn1r33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134169.1 | NP_598930.1 | 1 | 630 | ||
| GenBank mRNA | BC138428.1 | - | 1 | 679 | 8861448 | |
| BC138429.1 | - | 1 | 679 | 8861449 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r33| Gene Symbol: Vmn1r33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134169.1 | NP_598930.1 | 1 | 630 | ||
| GenBank mRNA | BC138428.1 | - | 1 | 679 | 8861448 | |
| BC138429.1 | - | 1 | 679 | 8861449 | ||