siRNA Details
| Gene Symbol: Vmn1r17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134171.1 | NP_598932.1 | 1 | 630 | ||
| Gene Symbol: Vmn1r31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166729.1 | NP_001160201.1 | 1 | 630 | ||

Vmn1r17
Genomic Map