siRNA Details
| Gene Symbol: Vmn1r5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134174.2 | NP_598935.2 | 1 | 575 | ||
| GenBank mRNA | BC138068.1 | - | 1 | 622 | 8861087 | |
| BC138069.1 | - | 1 | 622 | 8861088 | ||
| Gene Symbol: Vmn1r34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166719.1 | NP_001160191.1 | 1 | 575 | ||

Vmn1r5
Genomic Map