siRNA Details
| Gene Symbol: Vmn1r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134180.1 | NP_598941.1 | 1 | 429 | ||
| GenBank mRNA | AF394949.1 | - | 585 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r28| Gene Symbol: Vmn1r28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134180.1 | NP_598941.1 | 1 | 429 | ||
| GenBank mRNA | AF394949.1 | - | 585 | |||