siRNA Details
| Gene Symbol: Vmn1r8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134187.3 | NP_598948.1 | 1 | 278 | ||
| GenBank mRNA | AF394951.1 | - | 1 | 278 | ||
| Gene Symbol: Vmn1r7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166710.1 | NP_001160182.1 | 1 | 221 | ||

Vmn1r8
Genomic Map