siRNA Details
| Gene Symbol: Vmn1r198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134220.1 | NP_598981.1 | 1 | 31 | ||
| GenBank mRNA | BC130236.1 | - | 1 | 35 | 40142042 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r198| Gene Symbol: Vmn1r198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134220.1 | NP_598981.1 | 1 | 31 | ||
| GenBank mRNA | BC130236.1 | - | 1 | 35 | 40142042 | |