siRNA Details
| Gene Symbol: Vmn1r185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134231.1 | NP_598992.1 | 1 | 68 | ||
| GenBank mRNA | BC150720.1 | - | 1 | 154 | 9087631 | |
| Gene Symbol: Vmn1r184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167540.1 | NP_001161012.1 | 1 | 65 | ||

Vmn1r185
Genomic Map