siRNA Details
| Gene Symbol: Mela | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | D10049.1 | - | 915 | |||
| U63133.1 | - | 6422 | ||||
| Gene Symbol: Kcnma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | GU441834.1 | - | 1313 | |||

Mela
Genomic Map