siRNA Details
| Gene Symbol: Mip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008600.4 | NP_032626.2 | 3, 4 | 641 | ||
| GenBank mRNA | AK053490.1 | - | 3, 4 | 642 | ||
| AK053494.1 | - | 3, 4 | 604 | |||
| AK136287.1 | - | 3, 4 | 642 | |||
| BC082567.1 | - | 3, 4 | 630 | 30551292 | ||
| U27502.1 | - | 3, 4 | 597 | |||

Mip
Genomic Map