siRNA Details
| Gene Symbol: Mip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008600.4 | NP_032626.2 | 2, 3 | 567 | ||
| GenBank mRNA | AK053490.1 | - | 2, 3 | 568 | ||
| AK053494.1 | - | 2, 3 | 530 | |||
| AK136287.1 | - | 2, 3 | 568 | |||
| BC082567.1 | - | 2, 3 | 556 | 30551292 | ||
| U27502.1 | - | 2, 3 | 523 | |||

Mip
Genomic Map