siRNA Details
| Gene Symbol: Mpv17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294322.1 | NP_001281251.1 | 5 | 423 | ||
| NM_001294324.1 | NP_001281253.1 | 5 | 452 | |||
| NM_008622.6 | NP_032648.1 | 5 | 423 | |||
| GenBank mRNA | AK040477.1 | - | 5 | 393 | ||
| BC013452.1 | - | 4 | 376 | 4237284 | ||
| BY258057.1 | - | 5 | 395 | |||
| CK622051.1 | - | 4 | 365 | |||
| DV062332.1 | - | 5 | 505 | |||
| M36411.1 | - | 5 | 405 | |||

Mpv17
Genomic Map