siRNA Details
| Gene Symbol: Mpv17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294322.1 | NP_001281251.1 | 7 | 546 | ||
| NM_001294324.1 | NP_001281253.1 | 8 | 608 | |||
| NM_008622.6 | NP_032648.1 | 8 | 579 | |||
| GenBank mRNA | BC013452.1 | - | 7 | 532 | 4237284 | |
| CK622051.1 | - | 7 | 521 | |||
| DV062332.1 | - | 8 | 661 | |||
| DV660230.1 | - | 5 | 300 | |||
| M36411.1 | - | 8 | 561 | |||

Mpv17
Genomic Map