siRNA Details
| Gene Symbol: Mpv17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294322.1 | NP_001281251.1 | 4 | 325 | ||
| NM_001294324.1 | NP_001281253.1 | 4 | 354 | |||
| NM_008622.6 | NP_032648.1 | 4 | 325 | |||
| GenBank mRNA | AK040477.1 | - | 4 | 295 | ||
| BC013452.1 | - | 3 | 278 | 4237284 | ||
| CK622051.1 | - | 3 | 267 | |||
| DV062332.1 | - | 4 | 407 | |||
| DV660230.1 | - | 2 | 142 | |||
| M36411.1 | - | 4 | 307 | |||

Mpv17
Genomic Map