siRNA Details
Gene Symbol: Msi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008629.1 | NP_032655.1 | 6, 7 | 470 | ||
GenBank mRNA | AK084019.1 | - | 6, 7 | 453 | ||
D49654.1 | - | 6, 7 | 470 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008629.1 | NP_032655.1 | 6, 7 | 470 | ||
GenBank mRNA | AK084019.1 | - | 6, 7 | 453 | ||
D49654.1 | - | 6, 7 | 470 |