siRNA Details
| Gene Symbol: Mthfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161798.1 | NP_001155270.1 | 6 | 938 | ||
| NM_010840.3 | NP_034970.2 | 6 | 861 | |||
| GenBank mRNA | AK030192.1 | - | 6 | 856 | ||
| AK149795.1 | - | 6 | 924 | |||
| AK150159.1 | - | 6 | 939 | |||
| AK219632.1 | - | 3 | 249 | |||
| BC051017.1 | - | 6 | 840 | 6308248 | ||
| BC052466.1 | - | 6 | 905 | 6834886 | ||

Mthfr
Genomic Map