siRNA Details
| Gene Symbol: Mthfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161798.1 | NP_001155270.1 | 10, 11 | 1777 | ||
| NM_010840.3 | NP_034970.2 | 10, 11 | 1700 | |||
| GenBank mRNA | AK030192.1 | - | 10, 11 | 1695 | ||
| AK043144.1 | - | 2, 3 | 126 | |||
| AK149795.1 | - | 10, 11 | 1762 | |||
| AK150159.1 | - | 10, 11 | 1778 | |||
| BC051017.1 | - | 10, 11 | 1679 | 6308248 | ||
| BC052466.1 | - | 10, 11 | 1744 | 6834886 | ||
| BC100550.1 | - | 2, 3 | 168 | |||

Mthfr
Genomic Map