siRNA Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010855.3 | NP_034985.2 | 9, 10 | 903 | ||
| GenBank mRNA | AK138608.1 | - | 9, 10 | 904 | ||
| BC141362.1 | - | 7, 8 | 820 | 9055974 | ||
| BG796732.1 | - | 1, 2 | 144 | |||
| CR756940.1 | - | 3, 4 | 212 | |||

Myh4
Genomic Map