siRNA Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010855.3 | NP_034985.2 | 39 | 5736 | ||
| GenBank mRNA | AJ278733.1 | - | 9 | 1380 | ||
| AJ348869.1 | - | 2 | 151 | |||
| BC029240.1 | - | 6 | 774 | |||
| BC037757.1 | - | 4 | 422 | |||
| BC052786.1 | - | 4 | 430 | |||
| BC141362.1 | - | 37 | 5653 | 9055974 | ||
| K00988.1 | - | 2 | 157 | |||

Myh4
Genomic Map