siRNA Details
| Gene Symbol: Naga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008669.4 | NP_032695.3 | 3, 4 | 289 | ||
| GenBank mRNA | AF079458.1 | - | 3, 4 | 247 | ||
| AK077116.1 | - | 3, 4 | 261 | |||
| AK133576.1 | - | 3, 4 | 247 | |||
| AK153002.1 | - | 3, 4 | 275 | |||
| BC021631.1 | - | 3, 4 | 269 | 4019197 | ||
| CJ093426.1 | - | 3, 4 | 292 | |||

Naga
Genomic Map