siRNA Details
| Gene Symbol: Naga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008669.4 | NP_032695.3 | 9 | 1179 | ||
| GenBank mRNA | AF079458.1 | - | 9 | 1137 | ||
| AK077116.1 | - | 9 | 1151 | |||
| AK153002.1 | - | 9 | 1165 | |||
| BC021631.1 | - | 9 | 1159 | 4019197 | ||

Naga
Genomic Map