siRNA Details
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010877.4 | NP_035007.1 | 4, 5 | 643 | ||
GenBank mRNA | AB002664.1 | - | 4, 5 | 597 | ||
AK036379.1 | - | 4, 5 | 644 | |||
AK137152.1 | - | 4, 5 | 698 | |||
AK153447.1 | - | 4, 5 | 605 | |||
AK155324.1 | - | 4, 5 | 698 | |||
AK170839.1 | - | 4, 5 | 668 | |||
AK171059.1 | - | 4, 5 | 669 | |||
AK171265.1 | - | 4, 5 | 671 | |||
BC003730.1 | - | 4, 5 | 591 | 3592460 | ||
BC076609.1 | - | 5, 6 | 1597 | |||
BC094056.1 | - | 4, 5 | 688 | |||
DQ449939.1 | - | 4, 5 | 598 |
