siRNA Details
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010877.4 | NP_035007.1 | 7, 8 | 851 | ||
GenBank mRNA | AB002664.1 | - | 7, 8 | 805 | ||
AK036379.1 | - | 7, 8 | 852 | |||
AK137152.1 | - | 7, 8 | 906 | |||
AK153447.1 | - | 7, 8 | 813 | |||
AK155324.1 | - | 7, 8 | 906 | |||
AK170839.1 | - | 7, 8 | 876 | |||
AK171059.1 | - | 7, 8 | 877 | |||
AK171265.1 | - | 7, 8 | 879 | |||
BC003730.1 | - | 7, 8 | 799 | 3592460 | ||
BC076609.1 | - | 8, 9 | 1805 | |||
BC094056.1 | - | 7, 8 | 896 | |||
DQ449939.1 | - | 7, 8 | 806 |
