siRNA Details
| Gene Symbol: Nipsnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008698.2 | NP_032724.1 | 9 | 733 | ||
| GenBank mRNA | AJ001260.1 | - | 9 | 733 | ||
| AK031804.1 | - | 9 | 749 | |||
| BC010837.1 | - | 9 | 731 | 4242309 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nipsnap1| Gene Symbol: Nipsnap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008698.2 | NP_032724.1 | 9 | 733 | ||
| GenBank mRNA | AJ001260.1 | - | 9 | 733 | ||
| AK031804.1 | - | 9 | 749 | |||
| BC010837.1 | - | 9 | 731 | 4242309 | ||