siRNA Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008740.4 | NP_032766.2 | 10, 11 | 1194 | ||
| GenBank mRNA | AK049281.1 | - | 10, 11 | 1164 | ||
| AK085086.1 | - | 10, 11 | 1148 | |||
| AK153905.1 | - | 10, 11 | 1163 | |||
| AK216127.1 | - | 1, 2 | 97 | |||
| BC006627.1 | - | 10, 11 | 1199 | 3968733 | ||
| BC019167.1 | - | 10, 11 | 1147 | 4506351 | ||
| U10120.1 | - | 10, 11 | 1152 | |||

Nsf
Genomic Map