siRNA Details
| Gene Symbol: Oc90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010953.2 | NP_035083.1 | 11 | 851 | ||
| GenBank mRNA | AF091846.1 | - | 8 | 657 | ||
| AF093591.1 | - | 11 | 961 | |||
| AK012981.1 | - | 11 | 853 | |||
| BC048716.1 | - | 9 | 719 | 6334417 | ||

Oc90
Genomic Map