siRNA Details
Gene Symbol: Odf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008757.3 | NP_032783.2 | 2 | 516 | ||
GenBank mRNA | BC145758.1 | - | 2 | 516 | 40131061 | |
CJ042343.1 | - | 2 | 405 | |||
EU791469.1 | - | 2 | 516 | |||
X79446.1 | - | 2 | 520 |
