siRNA Details
| Gene Symbol: Olfr24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146606.1 | NP_666817.1 | 1 | 909 | ||
| GenBank mRNA | BC125621.1 | - | 1 | 985 | 40130136 | |
| BC137822.1 | - | 1 | 985 | 8860841 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr24| Gene Symbol: Olfr24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146606.1 | NP_666817.1 | 1 | 909 | ||
| GenBank mRNA | BC125621.1 | - | 1 | 985 | 40130136 | |
| BC137822.1 | - | 1 | 985 | 8860841 | ||