siRNA Details
| Gene Symbol: Olfr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146914.2 | NP_667125.1 | 2 | 378 | ||
| GenBank mRNA | BC104074.1 | - | 1 | 263 | ||
| BC104075.1 | - | 1 | 273 | 40041745 | ||
More Information
Set Membership: |
|

Olfr5
Genomic Map