siRNA Details
| Gene Symbol: Pbx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290576.1 | NP_001277505.1 | 4 | 819 | ||
| NM_016768.2 | NP_058048.1 | 4 | 819 | |||
| GenBank mRNA | AF020199.1 | - | 4 | 810 | ||
| AF020200.1 | - | 4 | 812 | |||
| AK144363.1 | - | 4 | 736 | |||
| BC057879.1 | - | 4 | 710 | 5348692 | ||

Pbx3
Genomic Map