siRNA Details
Gene Symbol: Cdk18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008795.2 | NP_032821.1 | 6, 7 | 812 | ||
GenBank mRNA | AK004998.1 | - | 6, 7 | 756 | ||
AK171569.1 | - | 6, 7 | 593 | |||
BC119617.1 | - | 7, 8 | 688 | 40061728 | ||
X69026.1 | - | 6, 7 | 829 |
