siRNA Details
| Gene Symbol: Pdcd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011051.3 | NP_035181.1 | 1, 2 | 215 | ||
| GenBank mRNA | AK008610.1 | - | 1, 2 | 139 | ||
| AK028223.1 | - | 1, 2 | 170 | |||
| AK078044.1 | - | 1, 2 | 143 | |||
| AK147979.1 | - | 1, 2 | 173 | |||
| AK150335.1 | - | 1, 2 | 133 | |||
| BC040079.1 | - | 1, 2 | 166 | 3156800 | ||
| BU604816.1 | - | 1, 2 | 218 | |||
| U49112.1 | - | 1, 2 | 152 | |||

Pdcd6
Genomic Map