siRNA Details
| Gene Symbol: Pex11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162387.1 | NP_001155859.1 | 4 | 670 | ||
| NM_001162388.1 | NP_001155860.1 | 4 | 434 | |||
| NM_011069.3 | NP_035199.3 | 4 | 670 | |||
| GenBank mRNA | AF093671.1 | - | 4 | 415 | ||
| AK005236.1 | - | 4 | 422 | |||
| AK011709.1 | - | 4 | 468 | |||
| AK082279.1 | - | 4 | 429 | |||
| AK082778.1 | - | 4 | 391 | |||
| BC013812.1 | - | 4 | 419 | 3483892 | ||
| BC054414.1 | - | 4 | 418 | 3257217 | ||

Pex11b
Genomic Map