siRNA Details
| Gene Symbol: Phf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011078.3 | NP_035208.2 | 10, 11 | 1350 | ||
| GenBank mRNA | AB221591.1 | - | 10, 11 | 1234 | ||
| AF043726.1 | - | 10, 11 | 1350 | |||
| AK153906.1 | - | 10, 11 | 1500 | |||
| AK172995.1 | - | 8, 9 | 1057 | |||
| BC051633.1 | - | 10, 11 | 1348 | 6518591 | ||

Phf2
Genomic Map