siRNA Details
| Gene Symbol: Pim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008842.3 | NP_032868.2 | 5 | 861 | ||
| GenBank mRNA | AK154127.1 | - | 204 | |||
| AK154669.1 | - | 4 | 700 | |||
| BC042885.1 | - | 5 | 1094 | 4507947 | ||
| BC053019.1 | - | 5 | 861 | 5694941 | ||
| BC055316.1 | - | 5 | 861 | 5694941 | ||

Pim1
Genomic Map