siRNA Details
| Gene Symbol: Pip5k1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293707.1 | NP_001280636.1 | 7 | 1250 | ||
| NM_008847.3 | NP_032873.2 | 7 | 1247 | |||
| GenBank mRNA | AK098097.1 | - | 7 | 1204 | ||
| AK150898.1 | - | 7 | 1207 | |||
| AK151984.1 | - | 7 | 1207 | |||
| AK158062.1 | - | 7 | 1192 | |||
| AK194373.1 | - | 1 | 108 | |||
| BC031774.1 | - | 7 | 1175 | 4503697 | ||

Pip5k1a
Genomic Map