siRNA Details
| Gene Symbol: Pip5k1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293707.1 | NP_001280636.1 | 13 | 1984 | ||
| NM_008847.3 | NP_032873.2 | 13 | 1981 | |||
| GenBank mRNA | AK098097.1 | - | 13 | 1938 | ||
| AK150898.1 | - | 13 | 1941 | |||
| AK151984.1 | - | 13 | 1941 | |||
| AK158062.1 | - | 13 | 1926 | |||
| BC003763.1 | - | 10 | 1038 | 3591003 | ||
| BC031774.1 | - | 13 | 1909 | 4503697 | ||
| D86177.1 | - | 13 | 1771 | |||

Pip5k1a
Genomic Map