siRNA Details
| Gene Symbol: Prkacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011100.4 | NP_035230.1 | 1, 2 | 315 | ||
| GenBank mRNA | AK048319.1 | - | 1, 2 | 307 | ||
| AK204631.1 | - | 1, 2 | 58 | |||
| AK216103.1 | - | 1, 2 | 110 | |||
| BC054533.1 | - | 1, 2 | 299 | 5704796 | ||
| J02626.1 | - | 1, 2 | 244 | |||

Prkacb
Genomic Map