siRNA Details
Gene Symbol: Pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011119.3 | NP_035249.1 | 9, 10 | 1050 | ||
GenBank mRNA | AK002685.1 | - | 9, 10 | 1127 | ||
AK076286.1 | - | 9, 10 | 1055 | |||
AK144645.1 | - | 9, 10 | 1104 | |||
AK166936.1 | - | 9, 10 | 1059 | |||
AK168582.1 | - | 9, 10 | 1052 | |||
BC003777.1 | - | 9, 10 | 1040 | |||
BC005593.1 | - | 9, 10 | 1040 | |||
BC028926.1 | - | 9, 10 | 1033 | |||
BC037623.1 | - | 9, 10 | 1070 | |||
BC046532.1 | - | 9, 10 | 1004 | 5358060 | ||
BC086788.1 | - | 7, 8 | 3256 | |||
U43918.1 | - | 9, 10 | 1209 | |||
X84789.1 | - | 9, 10 | 1115 |
