siRNA Details
| Gene Symbol: Pa2g4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011119.3 | NP_035249.1 | 10 | 1126 | ||
| GenBank mRNA | AK002685.1 | - | 10 | 1203 | ||
| AK076286.1 | - | 10 | 1132 | |||
| AK144645.1 | - | 10 | 1180 | |||
| AK166936.1 | - | 10 | 1135 | |||
| AK168582.1 | - | 10 | 1128 | |||
| BC003777.1 | - | 10 | 1116 | |||
| BC005593.1 | - | 10 | 1116 | |||
| BC028926.1 | - | 10 | 1109 | |||
| BC037623.1 | - | 10 | 1146 | |||
| BC046532.1 | - | 10 | 1080 | 5358060 | ||
| BC086788.1 | - | 8 | 3332 | |||
| U43918.1 | - | 10 | 1285 | |||
| X84789.1 | - | 10 | 1191 | |||

Pa2g4
Genomic Map