siRNA Details
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010127.3 | NP_034257.1 | 6 | 1599 | ||
GenBank mRNA | AB221558.1 | - | 5 | 889 | ||
AK006732.1 | - | 2 | 679 | |||
AY259046.1 | - | 10 | 2733 | |||
AY259047.1 | - | 9 | 2479 | |||
BC053046.1 | - | 8 | 1429 | |||
BC059830.1 | - | 6 | 1599 | 6410550 | ||
BC085139.1 | - | 11 | 1977 | 6406821 | ||
D13801.1 | - | 6 | 1088 | |||
L13763.1 | - | 5 | 922 |
