siRNA Details
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010127.3 | NP_034257.1 | 5 | 1135 | ||
GenBank mRNA | AB221558.1 | - | 4 | 425 | ||
AK006732.1 | - | 1 | 215 | |||
AY259046.1 | - | 9 | 2269 | |||
AY259047.1 | - | 8 | 2015 | |||
BC053046.1 | - | 7 | 965 | |||
BC059830.1 | - | 5 | 1135 | 6410550 | ||
BC085139.1 | - | 10 | 1513 | 6406821 | ||
D13801.1 | - | 5 | 624 | |||
L13763.1 | - | 4 | 458 |
