siRNA Details
Gene Symbol: Prl6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011166.2 | NP_035296.1 | 2 | 195 | ||
GenBank mRNA | AF011384.1 | - | 2 | 194 | ||
AF015563.1 | - | 2 | 195 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prl6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011166.2 | NP_035296.1 | 2 | 195 | ||
GenBank mRNA | AF011384.1 | - | 2 | 194 | ||
AF015563.1 | - | 2 | 195 |