siRNA Details
| Gene Symbol: Hps4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138646.3 | NP_619587.3 | 10, 11 | 1349 | ||
| GenBank mRNA | AK081489.1 | - | 10 | 1703 | ||
| AK129418.1 | - | 9, 10 | 1236 | |||
| AK147954.1 | - | 9, 10 | 1792 | |||
| AK171871.1 | - | 10, 11 | 1318 | |||
| AY043415.1 | - | 9, 10 | 869 | |||
| BC063055.1 | - | 9, 10 | 1041 | 5701206 | ||

Hps4
Genomic Map