siRNA Details
| Gene Symbol: Hps4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138646.3 | NP_619587.3 | 11 | 1711 | ||
| GenBank mRNA | AK081489.1 | - | 10 | 2065 | ||
| AK129418.1 | - | 10 | 1598 | |||
| AK147954.1 | - | 10 | 2154 | |||
| AK171871.1 | - | 11 | 1680 | |||
| AK178153.1 | - | 1 | 93 | |||
| AK178577.1 | - | 1 | 93 | |||
| AK179850.1 | - | 1 | 93 | |||
| AK202641.1 | - | 1 | 93 | |||
| AY043415.1 | - | 10 | 1231 | |||
| BC004668.1 | - | 1 | 318 | |||
| BC063055.1 | - | 10 | 1403 | 5701206 | ||

Hps4
Genomic Map