siRNA Details
| Gene Symbol: Hps4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138646.3 | NP_619587.3 | 11 | 1994 | ||
| GenBank mRNA | AK081489.1 | - | 10 | 2348 | ||
| AK129418.1 | - | 10 | 1881 | |||
| AK171871.1 | - | 11 | 1963 | |||
| AK185532.1 | - | 1 | 23 | |||
| AY043415.1 | - | 10 | 1514 | |||
| BC004668.1 | - | 1 | 601 | |||
| BC063055.1 | - | 10 | 1686 | 5701206 | ||

Hps4
Genomic Map